siRNA Details
| Gene Symbol: Zc3h18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201416.1 | NP_958819.1 | 8, 9 | 1542 | ||
| XM_006255758.2 | XP_006255820.1 | 9, 10 | 2205 | |||
| XM_006255759.2 | XP_006255821.1 | 9, 10 | 2206 | |||
| XM_006255760.2 | XP_006255822.1 | 8, 9 | 2132 | |||
| XM_006255761.2 | XP_006255823.1 | 9, 10 | 1716 | |||
| XM_008772603.1 | XP_008770825.1 | 9, 10 | 1624 | |||
| GenBank mRNA | AY389807.1 | - | 8, 9 | 1542 | ||
| AY389808.1 | - | 7, 8 | 1470 | |||

Zc3h18
Genomic Map