siRNA Details
| Gene Symbol: Rnaset2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106210.1 | NP_001099680.1 | 4 | 340 | ||
| GenBank mRNA | BC168957.1 | - | 373 | 9098746 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnaset2| Gene Symbol: Rnaset2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106210.1 | NP_001099680.1 | 4 | 340 | ||
| GenBank mRNA | BC168957.1 | - | 373 | 9098746 | ||