siRNA Details
| Gene Symbol: RGD1562747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106212.1 | NP_001099682.1 | 2, 3 | 515 | ||
| XM_006243225.2 | XP_006243287.1 | 2, 3 | 570 | |||
| GenBank mRNA | FQ213024.1 | - | 2, 3 | 490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562747| Gene Symbol: RGD1562747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106212.1 | NP_001099682.1 | 2, 3 | 515 | ||
| XM_006243225.2 | XP_006243287.1 | 2, 3 | 570 | |||
| GenBank mRNA | FQ213024.1 | - | 2, 3 | 490 | ||