siRNA Details
| Gene Symbol: Polb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017141.2 | NP_058837.2 | 12, 13 | 773 | ||
| GenBank mRNA | BC098668.1 | - | 12, 13 | 773 | 7386894 | |
| J02776.1 | - | 12, 13 | 761 | |||
| M13961.1 | - | 12, 13 | 751 | |||
| U38801.1 | - | 768 | ||||

Polb
Genomic Map