siRNA Details
| Gene Symbol: LOC292543 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025639.1 | NP_001020810.1 | 5 | 891 | ||
| XM_006228022.2 | XP_006228084.1 | 5 | 983 | |||
| XM_008758754.1 | XP_008756976.1 | 5 | 1056 | |||
| GenBank mRNA | BC098062.1 | - | 5 | 891 | 7458331 | |

LOC292543
Genomic Map