siRNA Details
| Gene Symbol: Vom2r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099472.1 | NP_001092942.1 | 2, 3 | 313 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom2r28| Gene Symbol: Vom2r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099472.1 | NP_001092942.1 | 2, 3 | 313 | ||
Set Membership: |
|