siRNA Details
| Gene Symbol: Nlrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169149.1 | NP_001162620.1 | 5 | 2206 | ||
| XM_006228188.1 | XP_006228250.1 | 4 | 2041 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp9| Gene Symbol: Nlrp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169149.1 | NP_001162620.1 | 5 | 2206 | ||
| XM_006228188.1 | XP_006228250.1 | 4 | 2041 | |||