siRNA Details
| Gene Symbol: Ceacam11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025404.1 | NP_001020575.1 | 3 | 635 | ||
| GenBank mRNA | BC097990.1 | - | 3 | 635 | 7445748 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ceacam11| Gene Symbol: Ceacam11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025404.1 | NP_001020575.1 | 3 | 635 | ||
| GenBank mRNA | BC097990.1 | - | 3 | 635 | 7445748 | |