siRNA Details
| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034920.2 | NP_001030092.1 | 7 | 1117 | ||
| GenBank mRNA | BC105776.1 | - | 7 | 1102 | 7931977 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cblc| Gene Symbol: Cblc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034920.2 | NP_001030092.1 | 7 | 1117 | ||
| GenBank mRNA | BC105776.1 | - | 7 | 1102 | 7931977 | |