siRNA Details
| Gene Symbol: Csrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017148.2 | NP_058844.1 | 4 | 423 | ||
| GenBank mRNA | BC062407.1 | - | 4 | 423 | 5623423 | |
| U09567.1 | - | 4 | 418 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csrp1| Gene Symbol: Csrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017148.2 | NP_058844.1 | 4 | 423 | ||
| GenBank mRNA | BC062407.1 | - | 4 | 423 | 5623423 | |
| U09567.1 | - | 4 | 418 | |||