siRNA Details
| Gene Symbol: Eif3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106242.1 | NP_001099712.1 | 3, 4 | 280 | ||
| XM_006228715.2 | XP_006228777.1 | 6, 7 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3k| Gene Symbol: Eif3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106242.1 | NP_001099712.1 | 3, 4 | 280 | ||
| XM_006228715.2 | XP_006228777.1 | 6, 7 | 587 | |||