siRNA Details
| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271398.1 | NP_001258327.1 | 12, 13 | 2274 | ||
| XM_008759099.1 | XP_008757321.1 | 13, 14 | 2230 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kirrel2| Gene Symbol: Kirrel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271398.1 | NP_001258327.1 | 12, 13 | 2274 | ||
| XM_008759099.1 | XP_008757321.1 | 13, 14 | 2230 | |||