siRNA Details
| Gene Symbol: Med25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170426.1 | NP_001163897.1 | 5 | 530 | ||
| XM_006229007.2 | XP_006229069.1 | 5 | 783 | |||
| XM_006229008.2 | XP_006229070.1 | 5 | 783 | |||
| XM_006229009.2 | XP_006229071.1 | 5 | 784 | |||
| XM_006229010.2 | XP_006229072.1 | 5 | 786 | |||
| XM_006229011.2 | XP_006229073.1 | 5 | 781 | |||
| XM_006229012.2 | XP_006229074.1 | 5 | 782 | |||
| XM_006229013.2 | XP_006229075.1 | 4 | 392 | |||

Med25
Genomic Map