siRNA Details
| Gene Symbol: Med25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170426.1 | NP_001163897.1 | 13 | 1458 | ||
| XM_006229007.2 | XP_006229069.1 | 13 | 1711 | |||
| XM_006229008.2 | XP_006229070.1 | 13 | 1708 | |||
| XM_006229009.2 | XP_006229071.1 | 13 | 1712 | |||
| XM_006229011.2 | XP_006229073.1 | 13 | 1706 | |||
| XM_006229012.2 | XP_006229074.1 | 13 | 1710 | |||
| XM_006229013.2 | XP_006229075.1 | 12 | 1320 | |||

Med25
Genomic Map