siRNA Details
| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039665.1 | NP_001034754.1 | 3 | 556 | ||
| XM_006229031.2 | XP_006229093.1 | 3 | 457 | |||
| XM_006229032.2 | XP_006229094.1 | 3 | 422 | |||
| XM_006229033.2 | XP_006229095.1 | 3 | 788 | |||
| XM_008759338.1 | XP_008757560.1 | 3 | 385 | |||
| GenBank mRNA | AY827147.1 | - | 3 | 556 | ||
| AY827148.1 | - | 3 | 425 | |||
| BC127438.1 | - | 1230 | ||||

Sult2b1
Genomic Map