siRNA Details
| Gene Symbol: Tsg101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181628.2 | NP_853659.2 | 8 | 801 | ||
| XM_008759341.1 | XP_008757563.1 | 6 | 702 | |||
| GenBank mRNA | AY293306.1 | - | 8 | 760 | ||
| BC070951.1 | - | 8 | 801 | 7100763 | ||
| FQ231102.1 | - | 5 | 481 | |||

Tsg101
Genomic Map