siRNA Details
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019185.1 | NP_062058.1 | 4, 5 | 1184 | ||
GenBank mRNA | L22760.1 | - | 4, 5 | 1184 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019185.1 | NP_062058.1 | 4, 5 | 1184 | ||
GenBank mRNA | L22760.1 | - | 4, 5 | 1184 |