siRNA Details
| Gene Symbol: Clec11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012459.1 | NP_001012477.1 | 2 | 311 | ||
| GenBank mRNA | AB009246.1 | - | 2 | 193 | ||
| BC090344.1 | - | 2 | 311 | 7304238 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clec11a| Gene Symbol: Clec11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012459.1 | NP_001012477.1 | 2 | 311 | ||
| GenBank mRNA | AB009246.1 | - | 2 | 193 | ||
| BC090344.1 | - | 2 | 311 | 7304238 | ||