siRNA Details
| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191601.1 | NP_001178530.1 | 5 | 614 | ||
| GenBank mRNA | FQ214686.1 | - | 5 | 616 | ||
| FQ232393.1 | - | 2 | 207 | |||
| FQ233613.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spcs2| Gene Symbol: Spcs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191601.1 | NP_001178530.1 | 5 | 614 | ||
| GenBank mRNA | FQ214686.1 | - | 5 | 616 | ||
| FQ232393.1 | - | 2 | 207 | |||
| FQ233613.1 | - | 2 | 207 | |||