siRNA Details
| Gene Symbol: Gpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183403.2 | NP_899653.2 | 1 | 227 | ||
| GenBank mRNA | BG664050.1 | - | 1 | 257 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpx2| Gene Symbol: Gpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183403.2 | NP_899653.2 | 1 | 227 | ||
| GenBank mRNA | BG664050.1 | - | 1 | 257 | ||