siRNA Details
Gene Symbol: Zfp36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017172.1 | NP_058868.1 | 1, 2 | 176 | ||
GenBank mRNA | X52590.1 | - | 1, 2 | 176 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp36l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017172.1 | NP_058868.1 | 1, 2 | 176 | ||
GenBank mRNA | X52590.1 | - | 1, 2 | 176 |