siRNA Details
| Gene Symbol: Ccdc101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114502.1 | NP_001107974.1 | 2, 3 | 269 | ||
| XM_006230231.2 | XP_006230293.1 | 2, 3 | 375 | |||
| XM_006230232.2 | XP_006230294.1 | 2, 3 | 279 | |||
| GenBank mRNA | FQ210727.1 | - | 2, 3 | 244 | ||
| FQ224509.1 | - | 2, 3 | 244 | |||
| FQ232716.1 | - | 2, 3 | 244 | |||

Ccdc101
Genomic Map