siRNA Details
| Gene Symbol: Slx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009292.1 | NP_001009292.1 | 4, 5 | 839 | ||
| GenBank mRNA | BC087090.1 | - | 4, 5 | 839 | 7190632 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slx1b| Gene Symbol: Slx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009292.1 | NP_001009292.1 | 4, 5 | 839 | ||
| GenBank mRNA | BC087090.1 | - | 4, 5 | 839 | 7190632 | |