siRNA Details
| Gene Symbol: Fgf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133286.1 | NP_579820.1 | 3, 4 | 227 | ||
| XM_006231436.2 | XP_006231498.1 | 3, 4 | 419 | |||
| XM_006231437.2 | XP_006231499.1 | 4, 5 | 347 | |||
| XM_008760398.1 | XP_008758620.1 | 4, 5 | 314 | |||
| GenBank mRNA | AB079113.1 | - | 3, 4 | 227 | ||

Fgf8
Genomic Map