siRNA Details
| Gene Symbol: Sox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019193.2 | NP_062066.1 | 5 | 1443 | ||
| GenBank mRNA | AJ001029.1 | - | 5 | 1444 | ||
| BC062067.1 | - | 4 | 1155 | 5599259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sox10| Gene Symbol: Sox10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019193.2 | NP_062066.1 | 5 | 1443 | ||
| GenBank mRNA | AJ001029.1 | - | 5 | 1444 | ||
| BC062067.1 | - | 4 | 1155 | 5599259 | ||