siRNA Details
Gene Symbol: Zdhhc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039100.1 | NP_001034189.1 | 3 | 856 | ||
GenBank mRNA | AY886540.1 | - | 3 | 856 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zdhhc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039100.1 | NP_001034189.1 | 3 | 856 | ||
GenBank mRNA | AY886540.1 | - | 3 | 856 |