siRNA Details
Gene Symbol: LOC100911841 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749038.3 | XP_003749086.1 | 5 | 520 |
Gene Symbol: Brms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009605.1 | NP_001009605.1 | 460 | |||
GenBank mRNA | BC088469.1 | - | 460 | 7128111 |
