siRNA Details
| Gene Symbol: Brms1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009605.1 | NP_001009605.1 | 403 | |||
| GenBank mRNA | BC088469.1 | - | 403 | 7128111 | ||
| Gene Symbol: LOC100911841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749038.3 | XP_003749086.1 | 4 | 463 | ||

Brms1
Genomic Map