siRNA Details
| Gene Symbol: Scyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011938.1 | NP_001011938.1 | 2 | 257 | ||
| XM_006230749.2 | XP_006230811.2 | 2 | 319 | |||
| GenBank mRNA | BC087141.1 | - | 2 | 257 | 7127562 | |
| FQ217187.1 | - | 2 | 307 | |||

Scyl1
Genomic Map