siRNA Details
| Gene Symbol: Tm7sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013071.1 | NP_001013089.1 | 7 | 838 | ||
| GenBank mRNA | BC091237.1 | - | 7 | 838 | 7323367 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm7sf2| Gene Symbol: Tm7sf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013071.1 | NP_001013089.1 | 7 | 838 | ||
| GenBank mRNA | BC091237.1 | - | 7 | 838 | 7323367 | |