siRNA Details
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011939.2 | NP_001011939.1 | 5 | 1379 | ||
GenBank mRNA | BC082030.1 | - | 5 | 1333 | ||
BC160908.1 | - | 5 | 1380 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ehd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011939.2 | NP_001011939.1 | 5 | 1379 | ||
GenBank mRNA | BC082030.1 | - | 5 | 1333 | ||
BC160908.1 | - | 5 | 1380 |