siRNA Details
| Gene Symbol: Mrpl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009647.1 | NP_001009647.1 | 4 | 633 | ||
| GenBank mRNA | BC058154.1 | - | 4 | 637 | ||
| BC088312.1 | - | 4 | 633 | 7324682 | ||
| EU148065.1 | - | 3 | 465 | |||
| FQ212609.1 | - | 4 | 643 | |||
| FQ214728.1 | - | 2 | 421 | |||
| FQ215871.1 | - | 4 | 645 | |||
| FQ219747.1 | - | 4 | 631 | |||

Mrpl16
Genomic Map