siRNA Details
| Gene Symbol: LOC100912050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749089.3 | XP_003749137.1 | 4 | 1031 | ||
| Gene Symbol: Dkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106350.1 | NP_001099820.1 | 844 | |||
| GenBank mRNA | JN966752.1 | - | 876 | |||

LOC100912050
Genomic Map