siRNA Details
Gene Symbol: Hoga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106355.1 | NP_001099825.1 | 2 | 508 | ||
GenBank mRNA | FQ210513.1 | - | 2 | 322 | ||
FQ218423.1 | - | 2 | 320 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hoga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106355.1 | NP_001099825.1 | 2 | 508 | ||
GenBank mRNA | FQ210513.1 | - | 2 | 322 | ||
FQ218423.1 | - | 2 | 320 |