siRNA Details
| Gene Symbol: Phf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212538.1 | NP_997703.1 | 15 | 1747 | ||
| XM_006256117.2 | XP_006256179.1 | 15 | 2001 | |||
| XM_008772723.1 | XP_008770945.1 | 15 | 2028 | |||
| GenBank mRNA | BC083843.1 | - | 15 | 1677 | 7114670 | |

Phf1
Genomic Map