siRNA Details
| Gene Symbol: Asf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106389.1 | NP_001099859.1 | 3 | 372 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Asf1a| Gene Symbol: Asf1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106389.1 | NP_001099859.1 | 3 | 372 | ||