siRNA Details
| Gene Symbol: Rab7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023950.3 | NP_076440.1 | 3 | 322 | ||
| XM_006236849.2 | XP_006236911.1 | 3 | 273 | |||
| GenBank mRNA | AB158427.1 | - | 2 | 154 | ||
| AB158428.1 | - | 2 | 154 | |||
| AF286535.1 | - | 3 | 318 | |||
| BC072470.1 | - | 3 | 322 | 7098657 | ||
| X12535.1 | - | 3 | 195 | |||

Rab7a
Genomic Map