siRNA Details
Gene Symbol: Pou5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009178.2 | NP_001009178.1 | 2 | 476 | ||
GenBank mRNA | BC158566.1 | - | 2 | 476 | 9092936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pou5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009178.2 | NP_001009178.1 | 2 | 476 | ||
GenBank mRNA | BC158566.1 | - | 2 | 476 | 9092936 |