siRNA Details
| Gene Symbol: Map6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017204.1 | NP_058900.1 | 3 | 2009 | ||
| XM_006229755.2 | XP_006229817.1 | 2 | 2339 | |||
| GenBank mRNA | AJ002556.1 | - | 2 | 2028 | ||
| BC078848.1 | - | 2 | 1887 | |||
| X93495.1 | - | 3 | 2009 | |||

Map6
Genomic Map