siRNA Details
| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017319.1 | NP_059015.1 | 3, 4 | 435 | ||
| GenBank mRNA | BC062393.1 | - | 3, 4 | 472 | 5621329 | |
| D63378.1 | - | 3, 4 | 435 | |||
| FQ211627.1 | - | 534 | ||||
| X12355.1 | - | 3, 4 | 431 | |||

Pdia3
Genomic Map