siRNA Details
| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017209.1 | NP_058905.1 | 1, 2 | 186 | ||
| GenBank mRNA | AB015724.1 | - | 1, 2 | 186 | ||
| FQ210707.1 | - | 1, 2 | 182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mecr| Gene Symbol: Mecr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017209.1 | NP_058905.1 | 1, 2 | 186 | ||
| GenBank mRNA | AB015724.1 | - | 1, 2 | 186 | ||
| FQ210707.1 | - | 1, 2 | 182 | |||