siRNA Details
| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013908.2 | NP_001013930.2 | 13 | 1492 | ||
| XM_006231884.2 | XP_006231946.1 | 13 | 1528 | |||
| XM_006231885.2 | XP_006231947.1 | 12 | 1513 | |||
| XM_006231886.2 | XP_006231948.1 | 12 | 1510 | |||
| GenBank mRNA | BC088172.1 | - | 13 | 1466 | 7323103 | |
| FQ217190.1 | - | 1475 | ||||

Trappc13
Genomic Map