siRNA Details
Gene Symbol: Rap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005765.1 | NP_001005765.1 | 5, 6 | 439 | ||
XM_006233076.1 | XP_006233138.1 | 3, 4 | 280 | |||
GenBank mRNA | BC083813.1 | - | 5, 6 | 439 | 7110254 | |
FQ233812.1 | - | 4, 5 | 345 |
