siRNA Details
| Gene Symbol: Rap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005765.1 | NP_001005765.1 | 6 | 465 | ||
| XM_006233076.1 | XP_006233138.1 | 4 | 306 | |||
| GenBank mRNA | BC083813.1 | - | 6 | 465 | 7110254 | |
| FQ233812.1 | - | 5 | 371 | |||

Rap1a
Genomic Map