siRNA Details
Gene Symbol: Hsd17b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017235.3 | NP_058931.1 | 5, 6 | 705 | ||
XM_006250215.2 | XP_006250277.1 | 5, 6 | 728 | |||
XM_006250216.2 | XP_006250278.1 | 5, 6 | 728 | |||
XM_006250217.2 | XP_006250279.1 | 5, 6 | 728 | |||
GenBank mRNA | FN803430.1 | - | 3, 4 | 318 | ||
FQ113345.1 | - | 5, 6 | 705 | |||
U44803.1 | - | 5, 6 | 650 |
