siRNA Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017237.3 | NP_058933.2 | 4 | 273 | ||
| GenBank mRNA | BC060573.1 | - | 4 | 277 | 6920680 | |
| D10699.1 | - | 4 | 257 | |||
| FQ214229.1 | - | 4 | 269 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uchl1| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017237.3 | NP_058933.2 | 4 | 273 | ||
| GenBank mRNA | BC060573.1 | - | 4 | 277 | 6920680 | |
| D10699.1 | - | 4 | 257 | |||
| FQ214229.1 | - | 4 | 269 | |||