siRNA Details
| Gene Symbol: Homer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053309.1 | NP_445761.1 | 4 | 594 | ||
| GenBank mRNA | AB007689.1 | - | 4 | 594 | ||
| AB007690.1 | - | 4 | 408 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Homer2| Gene Symbol: Homer2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053309.1 | NP_445761.1 | 4 | 594 | ||
| GenBank mRNA | AB007689.1 | - | 4 | 594 | ||
| AB007690.1 | - | 4 | 408 | |||