siRNA Details
| Gene Symbol: Rnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007641.1 | NP_001007642.1 | 1 | 230 | ||
| GenBank mRNA | AY461427.1 | - | 1 | 230 | ||
| BC168142.1 | - | 258 | 9100645 | |||
| FQ210593.1 | - | 269 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnd3| Gene Symbol: Rnd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007641.1 | NP_001007642.1 | 1 | 230 | ||
| GenBank mRNA | AY461427.1 | - | 1 | 230 | ||
| BC168142.1 | - | 258 | 9100645 | |||
| FQ210593.1 | - | 269 | ||||