siRNA Details
| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047857.3 | NP_001041322.2 | 3 | 594 | ||
| XM_006234405.2 | XP_006234467.1 | 3 | 521 | |||
| GenBank mRNA | DQ480743.1 | - | 3 | 597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2e3| Gene Symbol: Ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047857.3 | NP_001041322.2 | 3 | 594 | ||
| XM_006234405.2 | XP_006234467.1 | 3 | 521 | |||
| GenBank mRNA | DQ480743.1 | - | 3 | 597 | ||