siRNA Details
| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024755.1 | NP_001019926.1 | 3 | 706 | ||
| GenBank mRNA | BC097364.1 | - | 3 | 706 | 7443068 | |
| FQ220881.1 | - | 3 | 308 | |||
| FQ227712.1 | - | 3 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2l6| Gene Symbol: Ube2l6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024755.1 | NP_001019926.1 | 3 | 706 | ||
| GenBank mRNA | BC097364.1 | - | 3 | 706 | 7443068 | |
| FQ220881.1 | - | 3 | 308 | |||
| FQ227712.1 | - | 3 | 278 | |||